missing translation for 'onlineSavingsMsg'
Learn More

BAY1125976, MedChemExpress

Product Code. 16445487
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16445487 5 mg 5mg
16444206 100 mg 100mg
16429995 2 mg 2mg
16429446 50 mg 50mg
16422748 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16445487 Supplier Medchem Express Supplier No. HY100018
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BAY1125976 is a selective allosteric Akt1/Akt2 inhibitor; inhibits Akt1 and Akt2 activity with IC50 values of 5.2 nM and 18 nM at 10 μM ATP, respectively.

BAY1125976 is a selective allosteric Akt1/Akt2 inhibitor

Specifications

Chemical Name or Material BAY1125976
CAS 1402608-02-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H21N5O
Quantity 5 mg
Solubility Information DMSO : 25 mg/mL (65.20 mM; Need ultrasonic)
SMILES O=C(C1=NN2C(C=C1)=NC(C3=CC=C(C4(N)CCC4)C=C3)=C2C5=CC=CC=C5)N
Molecular Weight (g/mol) 383.45
Formula Weight 383.45
Percent Purity 99.74%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.