missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies (+)-Bay K8644
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
- (+)-Bay K8644, a modulator of potential operated calcium channels, exists as two enantiomers that have opposite actions (Hess et al.; Nowycky et al.; O'Neill et al.; Ravens et al.; Yamamoto et al.) (+)-Bay K8644 is an L-type channel blocker that has negative inotropic and vasodilatory effects (Artigas et al.; Franckowiak et al.; Ravens et al.)
- When combined with BIX-01294, enables reprogramming of mouse embryonic fibroblasts after transduction with OCT4 and KLF4 only (Shi et al.)
- Increases neuronal differentiation from neural stem and progenitor cells (NSCs) derived from the brains of postnatal mice (D'Ascenzo et al.)
Specifications
Specifications
| CAS | 98791-67-4 |
| Synonym | (+)-Bay-K 8644; (+)-Bay-R 4407; (R)-(+)-Bay K 8644; NI 105; R 4407 |
| Quantity | 10 mg |
| Molecular Formula | C16H15F3N2O4 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction