missing translation for 'onlineSavingsMsg'
Learn More

(±)-Bay K 8644, TRC

Product Code. 30132936
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132936 50 mg 50mg
30132423 100 mg 100mg
30006480 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132936 Supplier Toronto Research Chemicals Supplier No. TRCB11900850MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71145-03-4
Molecular Formula C16H15F3N2O4
Molecular Weight (g/mol) 356.3
Synonym (±)-Bay K 8644, Methyl 1,4-dihydro-2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-3-pyridinecarboxylate, BAY-k 8644, BK 8644, SQ 28873
IUPAC Name methyl 2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-1,4-dihydropyridine-3-carboxylate
SMILES COC(=O)C1=C(C)NC(=C(C1c2ccccc2C(F)(F)F)[N+](=O)[O-])C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H15F3N2O4/c1-8-12(15(22)25-3)13(14(21(23)24)9(2)20-8)10-6-4-5-7-11(10)16(17,18)19/h4-7,13,20H,1-3H3
Formula Weight 356.0984
Chemical Name or Material 1,4-Dihydro-2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-3-pyridinecarboxylic Acid, Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.