missing translation for 'onlineSavingsMsg'
Learn More

(±)-Bay K 8644, Thermo Scientific Chemicals

Product Code. 15434079
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15434079 1 mg 1mg
15424079 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15434079 Supplier Thermo Scientific Alfa Aesar Supplier No. J61480.M^

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

An active L-type, voltage-gated calcium channel agonist

An active L-type, voltage-gated calcium channel agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71145-03-4
Molecular Formula C16H15F3N2O4
Molecular Weight (g/mol) 356.301
MDL Number MFCD00036697
InChI Key ZFLWDHHVRRZMEI-UHFFFAOYSA-N
Synonym methyl 2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-1,4-dihydropyridine-3-carboxylate, 3-pyridinecarboxylic acid, 1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-, methyl ester, 1,4-dihydro-2,6-dimethyl-5-nitro-4-2′-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester, bay-k-8644, +--isomer, opera_id_789, d01rad, y-bay k 8644, r-+-1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester
PubChem CID 2303
ChEBI CHEBI:34555
IUPAC Name methyl 2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-1,4-dihydropyridine-3-carboxylate
SMILES CC1=C(C(C(=C(N1)C)[N+](=O)[O-])C2=CC=CC=C2C(F)(F)F)C(=O)OC

Specifications

Color Yellow
Quantity 1 mg
Solubility Information Soluble in DMSO, ethanol, or methanol; Insoluble in water
Formula Weight 356.3
Physical Form Powder
Chemical Name or Material (±)-Bay K 8644

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.