missing translation for 'onlineSavingsMsg'
Learn More

(±)-Bay K 8644, Thermo Scientific Chemicals

Product Code. 15424079
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 15424079

Brand: Thermo Scientific Alfa Aesar J61480.#0

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

An active L-type, voltage-gated calcium channel agonist

An active L-type, voltage-gated calcium channel agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71145-03-4
Molecular Formula C16H15F3N2O4
Molecular Weight (g/mol) 356.301
MDL Number MFCD00036697
InChI Key ZFLWDHHVRRZMEI-UHFFFAOYSA-N
Synonym methyl 2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-1,4-dihydropyridine-3-carboxylate, 3-pyridinecarboxylic acid, 1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-, methyl ester, 1,4-dihydro-2,6-dimethyl-5-nitro-4-2′-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester, bay-k-8644, +--isomer, opera_id_789, d01rad, y-bay k 8644, r-+-1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester
PubChem CID 2303
ChEBI CHEBI:34555
IUPAC Name methyl 2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-1,4-dihydropyridine-3-carboxylate
SMILES CC1=C(C(C(=C(N1)C)[N+](=O)[O-])C2=CC=CC=C2C(F)(F)F)C(=O)OC

Specifications

Color Yellow
Quantity 5 mg
Solubility Information Soluble in DMSO at 20mg/ml; Also soluble in methanol or ethanol at 60mg/ml; Insoluble in water
Formula Weight 356.3
Physical Form Powder
Chemical Name or Material (±)-Bay K 8644

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.