missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BAY 876
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
BAY 876 is a potent and selective GLUT1 inhibitor (IC50 = 2 nM) that displays selectivity for GLUT1 over GLUT2/3/4 (IC50 values are 10.8, 1.67 and 0.29 μM, respectively). BAY 876 induces cell death in hypoxic conditions in vitro and inhibits glucose uptake by Hela-MaTu cells. In ovarian cancer, BAY 876 reduces glycolysis rates and ATP production and inhibits proliferation in vitro and in vivo. This compound is cell permeable and orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | N4-[1-[(4-Cyanophenyl)methyl]-5-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-7-fluoro-2,4-quinolinedicarboxamide |
| CAS | 1799753-84-6 |
| Quantity | 25 mg |
| Molecular Formula | C24H16F4N6O2 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction