missing translation for 'onlineSavingsMsg'
Learn More

BAY-876, MedChemExpress

Product Code. p-200095832
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16435487

Brand: Medchem Express HY100017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAY-876 is an orally active and selective glucose transporter 1 (GLUT1) inhibitor with an IC50 of 2 nM. BAY-876 is >130-fold more selective for GLUT1 than GLUT2, GLUT3, and GLUT4. BAY-876 is also a potent blocker of glycolytic metabolism and ovarian cancer growth.

BAY-876 is an orally active and selective glucose transporter 1 (GLUT1) inhibitor with an IC50 of 2 nM. BAY-876 is >130-fold more selective for GLUT1 than GLUT2, GLUT3, and GLUT4. BAY-876 is also a potent blocker of glycolytic metabolism and ovarian cancer growth.

Specifications

Chemical Name or Material BAY-876
CAS 1799753-84-6
Color White
Purity Grade Notes Research
Molecular Formula C24H16F4N6O2
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (201.44 mM) ∣Methanol : 1 mg/mL (2.01 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C1=NC2=CC(F)=CC=C2C(C(NC3=C(C)N(CC4=CC=C(C#N)C=C4)N=C3C(F)(F)F)=O)=C1)N
Molecular Weight (g/mol) 496.42
Formula Weight 496.42
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.