missing translation for 'onlineSavingsMsg'
Learn More

Laduviglusib, MedChemExpress

Product Code. 16419996
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16419996

Brand: Medchem Express HY10182

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Laduviglusib (CHIR-99021) is a potent and selective GSK-3α/β inhibitor with IC50s of 10 nM and 6.7 nM. Laduviglusib shows >500-fold selectivity for GSK-3 over CDC2, ERK2 and other protein kinases. Laduviglusib is also a potent Wnt/β-catenin signaling pathway activator. Laduviglusib enhances mouse and human embryonic stem cells self-renewal. Laduviglusib induces autophagy.

Laduviglusib (CHIR-99021) is a potent and selective GSK-3α/β inhibitor with IC50s of 10 nM and 6.7 nM. Laduviglusib shows >500-fold selectivity for GSK-3 over CDC2, ERK2 and other protein kinases. Laduviglusib is also a potent Wnt/β-catenin signaling pathway activator. Laduviglusib enhances mouse and human embryonic stem cells self-renewal. Laduviglusib induces autophagy.

Specifications

Chemical Name or Material Laduviglusib
CAS 252917-06-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H18Cl2N8
Quantity 50 mg
Synonym CHIR-99021 CT99021
Solubility Information DMSO : 16.67 mg/mL (35.82 mM; Need ultrasonic)
SMILES N#CC1=CC=C(N=C1)NCCNC2=NC=C(C(C3=CC=C(Cl)C=C3Cl)=N2)C4=NC=C(N4)C
Molecular Weight (g/mol) 465.34
Formula Weight 465.34
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.