missing translation for 'onlineSavingsMsg'
Learn More

BAY-85-8501, MedChemExpress

Product Code. 16493706
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493706 25 mg 25mg
16487746 100 mg 100mg
16483887 50 mg 50mg
16467958 10 mg 10mg
16400428 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493706 Supplier Medchem Express Supplier No. HY19908
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress BAY-85-8501 is a selective, reversible and potent inhibitor of Human Neutrophil Elastase (HNE), with an IC50 of 65 pM.

BAY-85-8501 is a selective, reversible and potent inhibitor of Human Neutrophil Elastase (HNE), with an IC50 of 65 pM.

Specifications

Chemical Name or Material BAY-85-8501
CAS 1161921-82-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H17F3N4O3S
Quantity 25 mg
Solubility Information DMSO : 200 mg/mL (421.53 mM; Need ultrasonic)
SMILES N#CC1=C(C)N(C2=CC=CC(C(F)(F)F)=C2)C(N(C)[C@@H]1C3=CC=C(C#N)C=C3S(=O)(C)=O)=O
Molecular Weight (g/mol) 474.46
Formula Weight 474.46
Percent Purity 99.24%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.