missing translation for 'onlineSavingsMsg'
Learn More

BAY-8002, MedChemExpress

Product Code. 16497757
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497757 5 mg 5mg
16485776 100 mg 100mg
16435038 10 mg 10mg
16401277 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497757 Supplier Medchem Express Supplier No. HY122312
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress BAY-8002 is a potent, selective, orally active inhibitor of monocarboxylate transporter 1 (MCT1), with an IC50 of 85 nM in the MCT1-expressing DLD-1 cells, displays excellent selectivity against MCT4. Anti-tumor activity.

BAY-8002 is a potent, selective, orally active inhibitor of monocarboxylate transporter 1 (MCT1), with an IC50 of 85 nM in the MCT1-expressing DLD-1 cells, displays excellent selectivity against MCT4. Anti-tumor activity.

Specifications

Chemical Name or Material BAY-8002
CAS 724440-27-1
Color White
Purity Grade Notes Research
Molecular Formula C20H14ClNO5S
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (300.59 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=CC=C1NC(C2=CC(S(=O)(C3=CC=CC=C3)=O)=CC=C2Cl)=O
Molecular Weight (g/mol) 415.85
Formula Weight 415.85
Percent Purity 98.1%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.