missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BAY 678
GREENER_CHOICE

Product Code. 18741429
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18741429

Brand: Tocris Bioscience™ 5706/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective cell-permeable human neutrophil elastase (HNE) inhibitor

BAY 678 is a potent and selective cell-permeable human neutrophil elastase (HNE) inhibitor (IC50 = 20 nM). Exhibits around 2000-fold selectivity for NHE against a panel of 21 other serine proteases. Effective in an acute lung injury model in vivo. Orally bioavailable. Inactive S-enantiomer (Cat.No.6389) also available.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[(4R)-5-Acetyl-1,2,3,4-tetrahydro-6-methyl-2-oxo-1-[3-(trifluoromethyl)phenyl]-4-pyrimidinyl]-2-pyridinecarbonitrile
CAS 675103-36-3
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C20H15F3N4O2
Formula Weight Observed MW: 400.35
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.