missing translation for 'onlineSavingsMsg'
Learn More

BAY-677, MedChemExpress

Product Code. 16453096
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453096

Brand: Medchem Express HY111457

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAY-677 is an inactive control for BAY-678. BAY-678 is an orally bioavailable, highly potent, selective and cell-permeable inhibitor of human neutrophil elastase (HNE), with an IC50 of 20 nM. BAY-678 is also nominated as a chemical probe to the public via the Structural Genomics Consortium (SGC).

BAY-677 is an inactive control for BAY-678. BAY-678 is an orally bioavailable, highly potent, selective and cell-permeable inhibitor of human neutrophil elastase (HNE), with an IC50 of 20 nM. BAY-678 is also nominated as a chemical probe to the public via the Structural Genomics Consortium (SGC).

Specifications

Chemical Name or Material BAY-677
Physical Form Solid
CAS 2117404-84-7
Color Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C20H15F3N4O2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 50 mg/mL (124.89 mM)
SMILES N#CC1=NC=C([C@@H](C(C(C)=O)=C(C)N2C3=CC=CC(C(F)(F)F)=C3)NC2=O)C=C1
Molecular Weight (g/mol) 400.35
Formula Weight 400.35
Percent Purity 98.02%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.