missing translation for 'onlineSavingsMsg'
Learn More

Bay 41-4109 racemate, MedChemExpress

Product Code. 16454216
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16454216

Brand: Medchem Express HY100029A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAY 41-4109 racemate is the racemate of BAY 41-4109. BAY 41-4109 is a potent inhibitor of human hepatitis B virus (HBV) with an IC50 of 53 nM.

BAY 41-4109 racemate is the racemate of BAY 41-4109. BAY 41-4109 is a potent inhibitor of human hepatitis B virus (HBV) with an IC50 of 53 nM.

Specifications

Chemical Name or Material Bay 41-4109 racemate
CAS 298708-79-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H13ClF3N3O2
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (252.68 mM; Need ultrasonic)
SMILES O=C(C1=C(C)N=C(C2=NC=C(F)C=C2F)NC1C3=CC=C(F)C=C3Cl)OC
Molecular Weight (g/mol) 395.76
Formula Weight 395.76
Percent Purity 96.66%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.