missing translation for 'onlineSavingsMsg'
Learn More

BAY-2402234, MedChemExpress

Product Code. 16473186
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473186 25 mg 25mg
16470957 50 mg 50mg
16464668 10 mg 10mg
16455506 100 mg 100mg
16417387 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16473186 Supplier Medchem Express Supplier No. HY112645

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAY-2402234 is a selective dihydroorotate dehydrogenase (DHODH) inhibitor for the treatment of myeloid malignancies.

BAY-2402234 is a selective dihydroorotate dehydrogenase (DHODH) inhibitor for the treatment of myeloid malignancies.

Specifications

Chemical Name or Material BAY-2402234
CAS 2225819-06-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H18ClF5N4O4
Quantity 25 mg
Solubility Information DMSO : 125 mg/mL (240.00 mM; Need ultrasonic)
SMILES FC1=C(NC(C2=CC(F)=C(N3N=C(CO)N(CC)C3=O)C=C2O[C@@H](C)C(F)(F)F)=O)C(Cl)=CC=C1
Molecular Weight (g/mol) 520.84
Formula Weight 520.84
Percent Purity 98.71%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.