missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BAY 1816032
GREENER_CHOICE

Product Code. 18769776
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18769776

Brand: Tocris Bioscience™ 7549/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective BUB1 kinase inhibitor

BAY 1816032 is a potent and selective inhibitor of BUB1 kinase, a mitotic checkpoint serine/threonine kinase (IC50 = 6.1 nM). BAY 1816032 shows 17-fold selectivity over a panel of 403 other human kinases. In vivo, BAY 1816032 decreases tumor size in tumor xenograft models. BAY 181603 is orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[3,5-Difluoro-4-[[3-[5-methoxy-4-[(3-methoxy-4-pyridinyl)amino]-2-pyrimidinyl]-1 H-indazol-1-yl]methyl]phenoxy]ethanol
CAS 1891087-61-8
Quantity 10 mg
Target Checkpoint Control Kinase Inhibitors
Molecular Formula C27H24F2N6O4
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.