missing translation for 'onlineSavingsMsg'
Learn More

BAY 11-7085, MedChemExpress

Product Code. 16433748
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16433748

Brand: Medchem Express HY10257

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAY 11-7085 (BAY 11-7083) is an inhibitor of NF-κB activation and phosphorylation of IκBα; it stabilizes IκBα with an IC50 of 10 μM.

BAY 11-7085 (BAY 11-7083) is an inhibitor of NF-κB activation and phosphorylation of IκBα

Specifications

Chemical Name or Material BAY 11-7085
CAS 196309-76-9
Color White
Purity Grade Notes Research
Molecular Formula C13H15NO2S
Quantity 10 mg
Synonym BAY 11-7083
Solubility Information DMSO : ≥ 26 mg/mL (104.28 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES N#C/C=C/S(C(C=C1)=CC=C1C(C)(C)C)(=O)=O
Molecular Weight (g/mol) 249.33
Formula Weight 249.33
Percent Purity 99.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.