missing translation for 'onlineSavingsMsg'
Learn More

BAY 11-7082, MedChemExpress

Product Code. 16476186
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476186 100 mg 100mg
16468387 5 mg 5mg
16441857 50 mg 50mg
16425698 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476186 Supplier Medchem Express Supplier No. HY13453
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress BAY 11-7082 is an IκBα phosphorylation and NF-κB inhibitor.

BAY 11-7082 selectively and irreversibly inhibits the TNF-α-induced phosphorylation of IκB-α, and decreases NF-κB and expression of adhesion molecules. BAY 11-7082 inhibits ubiquitin-specific protease USP7 and USP21 (IC50=0.19, 0.96 μM, respectively). BAY 11-7082 inhibits gasdermin D (GSDMD) pore formation in liposomes and inflammasome-mediated pyroptosis and IL-1β secretion in human and mouse cells.

Specifications

Chemical Name or Material BAY 11-7082
CAS 19542-67-7
Color White
Purity Grade Notes Research
Molecular Formula C10H9NO2S
Quantity 100 mg
Synonym BAY 11-7821
Solubility Information DMSO : ≥ 100 mg/mL (482.51 mM)
SMILES N#C/C=C/S(=O)(C1=CC=C(C)C=C1)=O
Molecular Weight (g/mol) 207.25
Formula Weight 207.25
Percent Purity 99.28%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.