Learn More
Bax inhibitor 1 Antibody (20F430), mFluor Violet 500 SE, Novus Biologicals™
Product Code. 18424768
Product Code. 18424768 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP224919MFV500
Mouse Monoclonal Antibody
Bax inhibitor 1 Monoclonal antibody specifically detects Bax inhibitor 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)