missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bax inhibitor 1 Antibody (20F430) - Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-33264
This item is not returnable.
View return policy
Description
Bax inhibitor 1 Monoclonal specifically detects Bax inhibitor 1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Bax inhibitor 1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Immunohistochemistry-Paraffin 5-10 ug/ml | |
| BAXI1, BI1, BI-1Transmembrane BAX inhibitor motif-containing protein 6, TEGTbax inhibitor 1, testis enhanced gene transcript, Testis-enhanced gene transcript protein, transmembrane BAX inhibitor motif containing 6 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody is reactive to Human, Mouse, Rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| 20F430 | |
| Unconjugated | |
| P55061 | |
| TMBIM6 | |
| A mixture of three synthetic peptides corresponding to the N-terminal, internal and C-terminal regions of human B1. Epitope mapping indicates that the antibody recognizes an epitope in the N-terminal region of Bi-1. | |
| 0.1 mg | |
| Apoptosis | |
| 7009 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction