missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bax Antibody (BAX/8960R), Novus Biologicals™
Rabbit Monoclonal Antibody
£205.00 - £377.00
Specifications
| Antigen | Bax |
|---|---|
| Clone | BAX/8960R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18664035
|
Novus Biologicals
NBP3-23929-20ug |
20 μg |
£205.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679944
|
Novus Biologicals
NBP3-23929-100ug |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bax Monoclonal antibody specifically detects Bax in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Bax | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| 581 | |
| IgG κ | |
| Protein A or G purified |
| BAX/8960R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| apoptosis regulator BAX, BCL2-associated X protein, Bcl2-L-4, BCL2L4bcl2-L-4, Bcl-2-like protein 4 | |
| Recombinant full-length human Bax protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title