missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bavisant dihydrochloride hydrate, MedChemExpress
Description
Bavisant dihydrochloride hydrate (JNJ31001074AAC) is an orally active, potent, brain-penetrating and highly selective antagonist of the histamine H3 receptor. Bavisant dihydrochloride hydrate can be used for attention-deficit hyperactivity disorder (ADHD) research.
Specifications
Specifications
| Chemical Name or Material | Bavisant dihydrochloride hydrate |
| Physical Form | Solid |
| CAS | 1103522-80-0 |
| Color | Off-White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C19H31Cl2N3O3 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | JNJ31001074AACJNJ31001074 dihydrochloride hydrate |
| Solubility Information | H2O : ≥ 50 mg/mL (118.94 mM) ∣DMSO : ≥ 1 mg/mL (2.38 mM) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?