missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bavachin, MedChemExpress
Description
Bavachin, a flavonoid first isolated from seeds of P. corylifolia, acts as a phytoestrogen that activates the estrogen receptors ERα and ERβ with EC50s of 320 and 680 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Bavachin |
| CAS | 19879-32-4 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C20H20O4 |
| Quantity | 5 mg |
| Synonym | Corylifolin |
| Solubility Information | DMSO : 100 mg/mL (308.29 mM; Need ultrasonic) |
| SMILES | O=C1C[C@@H](C2=CC=C(O)C=C2)OC3=CC(O)=C(C/C=C(C)\C)C=C13 |
| Molecular Weight (g/mol) | 324.37 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?