missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ BATTERY STORE Storage Cabinet ION-CLASSIC-90

Safe passive storage of lithium-ion batteries

£2827.55 - £3928.89

Specifications

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, CE compliant
Color Anthracite Gray, Gentian Blue
Material Sheet Steel
Product Type Storage cabinet
Door Style Wing
View More Specs

Alert:

For indoor use only


Products 8
Product Code Brand No. of Doors No. of Shelves Price Quantity & Availability  
Product Code Brand No. of Doors No. of Shelves Price Quantity & Availability  
16359426
View Documents Promotion Details
Asecos™
3806704738086
1 3 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16369426
View Documents Promotion Details
Asecos™
3806704738087
1 4 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16379426
View Documents Promotion Details
Asecos™
3806704738088
1 5 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16389426
View Documents Promotion Details
Asecos™
3806704738089
1 6 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16379406
View Documents Promotion Details
Asecos™
3725404737258
2 3 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16389406
View Documents Promotion Details
Asecos™
3725404737264
2 4 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16399406
View Documents Promotion Details
Asecos™
3725404737265
2 5 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16309416
View Documents Promotion Details
Asecos™
3725404737266
2 6 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Type 90 Safety storage cabinet
  • Tall cabinet with self-closing wing doors
  • With cylinder locking and locking state indicator
  • Integrated adjusting aids to compensate uneven floor
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
  • Safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Integrated transport base with removable, optional base cover
  • Fire resistance 90 minutes
  • Type tested in accordance with EN 14470-1
  • CE compliant
Specifications

Specifications

Fire resistance 90 minutes, type tested in accordance with EN 14470-1, CE compliant
Sheet Steel
Wing
Bottom collecting sump (sheet steel powder-coated)
Powder-coated textured
Anthracite Gray, Gentian Blue
Storage cabinet
Safe passive storage of lithium-ion batteries
Grid
Videos
SDS
Documents

Documents

Product Certifications