missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ BATTERY STORE Storage Cabinet ION-CLASSIC-90

Product Code. 16379406 Shop All asecos Products
Click to view available options
No. of Doors:
1
2
No. of Shelves:
3
4
5
6
Unit Size:
Each
This item is not returnable. View return policy

For indoor use only

Product Code. 16379406

Brand: Asecos™ 3725404737258

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For indoor use only

Safe passive storage of lithium-ion batteries

  • Type 90 Safety storage cabinet
  • Tall cabinet with self-closing wing doors
  • With cylinder locking and locking state indicator
  • Integrated adjusting aids to compensate uneven floor
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
  • Safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Integrated transport base with removable, optional base cover
  • Fire resistance 90 minutes
  • Type tested in accordance with EN 14470-1
  • CE compliant

Especificaciones

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, CE compliant
Dimensions (D x W x H) Interior 522 x 1050 x 1647 mm
Dimensions (D x W x H) 615 x 1193 x 2224 mm
Door Style Wing
For Use With (Application) Safe passive storage of lithium-ion batteries
Includes Bottom collecting sump (sheet steel powder-coated)
Model IO90.195.120.K1.WDC
No. of Doors 2
No. of Shelves 3
Shelf Type Grid
Surface Coating Powder-coated textured
Color Anthracite Gray, Gentian Blue
Material Sheet Steel
Product Type Storage cabinet
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado