missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Batimastat, Tocris Bioscience™
Potent, broad spectrum MMP inhibitor
£149.40 - £302.12
Chemical Identifiers
CAS | 130370-60-4 |
---|---|
Molecular Formula | C23H31N3O4S2 |
Molecular Weight (g/mol) | 477.638 |
InChI Key | XFILPEOLDIKJHX-QYZOEREBSA-N |
Synonym | batimastat, batimastat bb-94, unii-bk349f52c9, butanediamide, n4-hydroxy-n1-2-methylamino-2-oxo-1-phenylmethyl ethyl-2-2-methylpropyl-3-2-thienylthio methyl-, 2r-1 s* ,2r*,3s*, 2s,3r-5-methyl-3-alphas-alpha-methylcarbamoyl phenethyl carbamoyl-2-2-thienylthio methyl hexanohydroxamic acid, 4-n-hydroxyamino-2r-isobutyl-2s-2-thienylthiomethyl succinyl-l-phenylalanine-n-methylamide, 2r-n-1s-1-benzyl-2-methylamino-2-oxo-ethyl-2-1s-2-hydroxyamino-2-oxo-1-2-thienylsulfanylmethyl ethyl-4-methyl-pentanamide, 2r,3s-n-hydroxy-n'-1s-1-methylcarbamoyl-2-phenylethyl-2-2-methylpropyl-3-thiophen-2-ylsulfanyl methyl butanediamide, butanediamide, n sup 4-hydroxy-n sup 1-2-methylamino-2-oxo-1-phenylmethyl ethyl-2-2-methylpropyl-3-2-thienylthio methyl-, 2r-1 s* ,2r*,3s* |
PubChem CID | 5362422 |
IUPAC Name | (2S,3R)-N-hydroxy-N'-[(2S)-1-(methylamino)-1-oxo-3-phenylpropan-2-yl]-3-(2-methylpropyl)-2-(thiophen-2-ylsulfanylmethyl)butanediamide |
SMILES | CC(C)CC(C(CSC1=CC=CS1)C(=O)NO)C(=O)NC(CC2=CC=CC=C2)C(=O)NC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18063792
|
Tocris Bioscience
2961/1 |
1 mg |
£149.40
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18053962
|
Tocris Bioscience
2961/10 |
10 mg |
£302.12
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent, broad spectrum MMP inhibitor (IC50 values are 3, 4, 4, 6 and 20 nM for MMP -1, -2, -9, -7 and -3 respectively). Exhibits antiproliferative, anti-invasive and antimetastatic activity in human ovarian carcinoma xenografts in vivo.Chemical Identifiers
130370-60-4 | |
477.638 | |
batimastat, batimastat bb-94, unii-bk349f52c9, butanediamide, n4-hydroxy-n1-2-methylamino-2-oxo-1-phenylmethyl ethyl-2-2-methylpropyl-3-2-thienylthio methyl-, 2r-1 s* ,2r*,3s*, 2s,3r-5-methyl-3-alphas-alpha-methylcarbamoyl phenethyl carbamoyl-2-2-thienylthio methyl hexanohydroxamic acid, 4-n-hydroxyamino-2r-isobutyl-2s-2-thienylthiomethyl succinyl-l-phenylalanine-n-methylamide, 2r-n-1s-1-benzyl-2-methylamino-2-oxo-ethyl-2-1s-2-hydroxyamino-2-oxo-1-2-thienylsulfanylmethyl ethyl-4-methyl-pentanamide, 2r,3s-n-hydroxy-n'-1s-1-methylcarbamoyl-2-phenylethyl-2-2-methylpropyl-3-thiophen-2-ylsulfanyl methyl butanediamide, butanediamide, n sup 4-hydroxy-n sup 1-2-methylamino-2-oxo-1-phenylmethyl ethyl-2-2-methylpropyl-3-2-thienylthio methyl-, 2r-1 s* ,2r*,3s* | |
(2S,3R)-N-hydroxy-N'-[(2S)-1-(methylamino)-1-oxo-3-phenylpropan-2-yl]-3-(2-methylpropyl)-2-(thiophen-2-ylsulfanylmethyl)butanediamide |
C23H31N3O4S2 | |
XFILPEOLDIKJHX-QYZOEREBSA-N | |
5362422 | |
CC(C)CC(C(CSC1=CC=CS1)C(=O)NO)C(=O)NC(CC2=CC=CC=C2)C(=O)NC |
Specifications
130370-60-4 | |
batimastat, batimastat bb-94, unii-bk349f52c9, butanediamide, n4-hydroxy-n1-2-methylamino-2-oxo-1-phenylmethyl ethyl-2-2-methylpropyl-3-2-thienylthio methyl-, 2r-1 s* ,2r*,3s*, 2s,3r-5-methyl-3-alphas-alpha-methylcarbamoyl phenethyl carbamoyl-2-2-thienylthio methyl hexanohydroxamic acid, 4-n-hydroxyamino-2r-isobutyl-2s-2-thienylthiomethyl succinyl-l-phenylalanine-n-methylamide, 2r-n-1s-1-benzyl-2-methylamino-2-oxo-ethyl-2-1s-2-hydroxyamino-2-oxo-1-2-thienylsulfanylmethyl ethyl-4-methyl-pentanamide, 2r,3s-n-hydroxy-n'-1s-1-methylcarbamoyl-2-phenylethyl-2-2-methylpropyl-3-thiophen-2-ylsulfanyl methyl butanediamide, butanediamide, n sup 4-hydroxy-n sup 1-2-methylamino-2-oxo-1-phenylmethyl ethyl-2-2-methylpropyl-3-2-thienylthio methyl-, 2r-1 s* ,2r*,3s* | |
CC(C)CC(C(CSC1=CC=CS1)C(=O)NO)C(=O)NC(CC2=CC=CC=C2)C(=O)NC | |
477.638 | |
477.64 | |
Batimastat |
C23H31N3O4S2 | |
XFILPEOLDIKJHX-QYZOEREBSA-N | |
(2S,3R)-N-hydroxy-N'-[(2S)-1-(methylamino)-1-oxo-3-phenylpropan-2-yl]-3-(2-methylpropyl)-2-(thiophen-2-ylsulfanylmethyl)butanediamide | |
5362422 | |
>99% |
Safety and Handling
Recommended Storage : Store at -20°C