missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JULABO™ Bath Fluid Thermal HL60
Description
- JULABO Thermal bath fluids have been carefully selected and are optimized for maximum performance with JULABO instruments
- Low viscosity
- High stability
- Minimum odor
- Good heat conductivity
- Wide temperature ranges
- Including drain tap
Specifications
Specifications
| Capacity (Metric) | 10 L |
| For Use With (Equipment) | PRESTO™ A30, PRESTO™ A45, PRESTO™ A45t, PRESTO™ A80, PRESTO™ A80t, PRESTO™ A85, PRESTO™ A85t, PRESTO™ W40, PRESTO™ W50, PRESTO™ W50t, PRESTO™ W80, PRESTO™ W80t, PRESTO™ W85, PRESTO™ W85t, PRESTO™ W91, PRESTO™ W91tt, PRESTO™ W91x, PRESTO™ W92, PRESTO™ W92tt, PRESTO™ W92ttx, PRESTO™ W92x |
| Temperature (Metric) Maximum | -60°C to +250°C |
| Viscosity | 5.66 (kinematic at 20 °C in mm²/s) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?