Learn More
HUBER Bath Cover
Product Code. 10101135
Change view
Click to view available options
For Use With (Equipment):
11 L Variostat Tank
22 L Variostat Tank
Unical 700, CAL 1-4, Unical 1
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10101135 Unical 700, CAL 1-4, Unical 1 Each
10282835 11 L Variostat Tank Each
10606527 22 L Variostat Tank Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10101135 Supplier HUBER Supplier No. 6367
Bath covers, accessories for Huber temperature control units
Bath cover, insulated with opening
Specifications
For Use With (Equipment) Unical 700, CAL 1-4, Unical 1
Temperature Range -100°C to +300°C