Learn More
Nickel Electro™ Bath Armor
Product Code. 12724866 Shop All Nickel-Electro Products
Change view
Click to view available options
Material:
Aluminum
Unit Size:
12 L
Product Code. Material unitSize
12724866 Aluminum 12 L
1 options
Product Code. 12724866 Supplier Nickel Electro™ Supplier No. BA12
Only null left
Add to basket
Add to basket
Designed as a replacement to water in a bath. Nickel Electro™ Bath Armor is thermally efficient aluminum beads that elimnites evaporation and run dry issues.
  • No more filling topping up water baths, maintaining water level
  • Cleaning frequency reduced
  • No water bourne contamination
  • Good thermal heat distribution
  • Replace water in your water bath with these thermally efficient pellets that are easy to keep clean
  • It enables you to eliminate nearly all water bath cleaning and protects your water bath from drying out
  • Saving the need for racks, floaties and bottle weights
  • Aims to tackle water contamination by utilizing a dry, metallic thermal media comprised of small beads instead of water
  • The result is a dry bath that is far less conducive to contamination than a water-filled bath
  • Do not autoclave
Specifications
Description Bath Armor
Type Bath Armor
Material Aluminum
Quantity 12 L
Shape Round