Learn More
BATF3 Antibody (841702) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB7437E
Product Code. 18573165
  • £516.00 / 0.10mL
Description
BATF3 Monoclonal antibody specifically detects BATF3 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BATF3
Monoclonal
DyLight 405
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry (Intracellular Staining), CyTOF
841702
Intracellular Staining by Flow Cytometry, CyTOF-ready
basic leucine zipper transcription factor, ATF-like 3, basic leucine zipper transcriptional factor ATF-like 3, B-ATF-3, FLJ36352, FLJ37535,21 kDa small nuclear factor isolated from T-cells, JDP1, Jun dimerization protein 1, Jun dimerization protein p21SNFT, JUNDM1, SNFT21-kD small nuclear factor isolated from T cells
E. coli-derived recombinant human BATF3, Met1-Arg127, Accession # Q9NR55
0.1 mL
Immunology
55509
Store at 4°C in the dark.
IgG2b