missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BATF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564028
This item is not returnable.
View return policy
Description
Immunogen sequence: MHLCGGNGLL TQTDPKEQQR QLKKQKNRAA A Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 66%.
BATF2 (basic leucine zipper transcription factor, ATF-like 2) is a 274 amino acid protein that localizes to the nucleus and contains one bZIP domain, suggesting that it may be involved in transcriptional regulation. The gene encoding BATF2, which is expressed as multiple alternatively spliced isoforms, is located on human chromosome 11. With approximately 135 million base pairs and 1, 400 genes, chromosome 11 comprises approximately 4% of human genomic DNA and is considered a gene and disease association dense chromosome. The chromosome 11 encoded Atm gene is important for regulation of cell cycle arrest and apoptosis following double strand DNA breaks. Atm mutation leads to the disorder known as ataxia-telangiectasia. The blood disorders Sickle cell anemia and thalassemia are caused by HBB gene mutations, while Wilms' tumors, WAGR syndrome and Denys-Drash syndrome are associated with mutations of the WT1 gene. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are also associated with defects in chromosome 11-encoded genes.
Specifications
| BATF2 | |
| Polyclonal | |
| Unconjugated | |
| BATF2 | |
| 4933430F08Rik; basic leucine zipper ATF-like transcription factor 2; basic leucine zipper transcription factor, ATF-like 2; basic leucine zipper transcriptional factor ATF-like 2; BATF2; B-ATF-2; MGC20410; RGD1311481; SARI; Suppressor of AP-1 regulated by IFN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116071 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N1L9 | |
| BATF2 | |
| Recombinant protein corresponding to Human BATF2. Recombinant protein control fragment (Product #RP-103454). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction