missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BATF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13507-1-AP-20UL
This item is not returnable.
View return policy
Description
BATF is a nuclear basic leucine zipper protein that belongs to the AP-1/ATF superfamily of transcription factors. The leucine zipper of this protein mediates dimerization with members of the Jun family of proteins. This protein is thought to be a negative regulator of AP-1/ATF transcriptional events and blocks cellular transformation by Ras and Fos. BATF also is required for the differentiation of IL-17-producing T helper (TH17) cells, a CD4+ subset of T cells that coordinates the inflammatory response in host defense. In both T cells and B cells, BATF is required for the appropriate regulation of the class-switch recombination.Specifications
| BATF | |
| Polyclonal | |
| Unconjugated | |
| BATF | |
| B ATF, BATF, BATF1, SF HT activated gene 2 protein, SFA 2, SFA2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10538, 299206, 53314 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| D4A7E1, O35284, Q16520 | |
| Batf | |
| BATF Fusion Protein Ag4418 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction