missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BATF Polyclonal Antibody
GREENER_CHOICE

Product Code. 17199403
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17199403 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17199403 Supplier Invitrogen™ Supplier No. PA5111491

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DSSDSSFSRS PPPGKQDSSD DVRRVQRREK Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.

BATF is a nuclear basic leucine zipper protein that belongs to the AP-1/ATF superfamily of transcription factors. The leucine zipper of this protein mediates dimerization with members of the Jun family of proteins. This protein is thought to be a negative regulator of AP-1/ATF transcriptional events and blocks cellular transformation by Ras and Fos. BATF also is required for the differentiation of IL-17-producing T helper (TH17) cells, a CD4+ subset of T cells that coordinates the inflammatory response in host defense. In both T cells and B cells, BATF is required for the appropriate regulation of the class-switch recombination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BATF
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Batf
Gene Accession No. Q16520
Gene Alias activating transcription factor B; basic leucine zipper ATF-like transcription factor; basic leucine zipper transcription factor, ATF-like; basic leucine zipper transcription factor-like; basic leucine zipper transcriptional factor ATF-like; BATF; B-ATF; BATF1; B-cell activating transcription factor; B-cell-activating transcription factor; SFA2; SFA-2; SF-HT-activated gene 2 protein; si:ch211-153j24.4
Gene Symbols Batf
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human BATF. Recombinant protein control fragment (Product #RP-106950).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10538
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.