missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BATF Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534463
This item is not returnable.
View return policy
Description
A suggested positive control is rat spleen tissue lysate. PA5-34463 can be used with blocking peptide PEP-1505.
BATF is a nuclear basic leucine zipper protein that belongs to the AP-1/ATF superfamily of transcription factors. The leucine zipper of this protein mediates dimerization with members of the Jun family of proteins. This protein is thought to be a negative regulator of AP-1/ATF transcriptional events and blocks cellular transformation by Ras and Fos. BATF also is required for the differentiation of IL-17-producing T helper (TH17) cells, a CD4+ subset of T cells that coordinates the inflammatory response in host defense. In both T cells and B cells, BATF is required for the appropriate regulation of the class-switch recombination.
Specifications
| BATF | |
| Polyclonal | |
| Unconjugated | |
| Batf | |
| activating transcription factor B; basic leucine zipper ATF-like transcription factor; basic leucine zipper transcription factor, ATF-like; basic leucine zipper transcription factor-like; basic leucine zipper transcriptional factor ATF-like; BATF; B-ATF; BATF1; B-cell activating transcription factor; B-cell-activating transcription factor; SFA2; SFA-2; SF-HT-activated gene 2 protein; si:ch211-153j24.4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10538, 299206, 53314 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| D4A7E1, O35284, Q16520 | |
| Batf | |
| A 16 amino acid synthetic peptide near the amino terminus of human BATF. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction