missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BATF Mouse anti-Human, Clone:7C8F3, Abnova™
Mouse monoclonal antibody raised against full length recombinant human BATF.
Brand: Abnova MAB17950.100ug
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a nuclear basic leucine zipper protein that belongs to the AP-1/ATF superfamily of transcription factors. The leucine zipper of this protein mediates dimerization with members of the Jun family of proteins. This protein is thought to be a negative regulator of AP-1/ATF transcriptional events. [provided by RefSeq]Specifications
| BATF | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
| BATF | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 7C8F3 | |
| basic leucine zipper transcription factor, ATF-like | |
| In PBS (0.05% sodium azide) | |
| B-ATF, BATF1, SFA-2, SFA2 | |
| Recombinant protein corresponding to full length human BATF. | |
| 10538 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction