missing translation for 'onlineSavingsMsg'
Learn More

BATF Antibody (8A12), Novus Biologicals™

Product Code. 18380239 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380239 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18380239 Supplier Novus Biologicals Supplier No. H00010538M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BATF Monoclonal antibody specifically detects BATF in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation, ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BATF
Applications Western Blot, ELISA, Immunoprecipitation, ChIP Assay
Classification Monoclonal
Clone 8A12
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP)
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_006390
Gene Alias basic leucine zipper transcription factor, ATF-like, BATF1basic leucine zipper transcriptional factor ATF-like, B-ATFSFA2, B-cell-activating transcription factor, SFA-2activating transcription factor B, SF-HT-activated gene 2 protein
Host Species Mouse
Immunogen BATF (NP_006390, 34 a.a. ~ 125 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EKNRIAAQKSRQRQTQKADTLHLESEDLEKQNAALRKEIKQLTEELKYFTSVLNSHEPLCSVLAASTPSPPEVVYSAHAFHQPHVSSPRFQP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10538
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.