missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BATF Antibody (687706) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8054JF635
This item is not returnable.
View return policy
Description
BATF Monoclonal antibody specifically detects BATF in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| BATF | |
| Monoclonal | |
| Janelia Fluor 635 | |
| basic leucine zipper transcription factor, ATF-like, BATF1basic leucine zipper transcriptional factor ATF-like, B-ATFSFA2, B-cell-activating transcription factor, SFA-2activating transcription factor B, SF-HT-activated gene 2 protein | |
| E. coli-derived recombinant human BATF , Met1-Pro125, Accession # Q16520 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Western Blot, CyTOF | |
| 687706 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Signal Transduction | |
| 10538 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction