missing translation for 'onlineSavingsMsg'
Learn More

Batefenterol, MedChemExpress

Product Code. 16495458
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495458 10 mg 10mg
16461637 50 mg 50mg
16438177 5 mg 5mg
16426056 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495458 Supplier Medchem Express Supplier No. HY12980

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Batefenterol (GSK961081;TD-5959) is a novel muscarinic receptor antagonist and β2-adrenoceptor agonist; displays high affinity for hM2, hM3 muscarinic and hβ2-adrenoceptor with Ki values of 1.4, 1.3 and 3.7 nM, respectively.

Batefenterol (GSK961081

Specifications

Chemical Name or Material Batefenterol
CAS 743461-65-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C40H42ClN5O7
Quantity 10 mg
Synonym GSK961081 TD-5959
Solubility Information DMSO : ≥ 100 mg/mL (135.09 mM)
SMILES O=C(OC1CCN(CCC(NC2=CC(OC)=C(CNC[C@H](O)C3=CC=C(O)C4=C3C=CC(N4)=O)C=C2Cl)=O)CC1)NC5=CC=CC=C5C6=CC=CC=C6
Molecular Weight (g/mol) 740.24
Formula Weight 740.24
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.