missing translation for 'onlineSavingsMsg'
Learn More

Brannan™ Batch Sampling Oven Thermometer with Calibration Certificate

Product Code. 17873288
Change view
Click to view available options
Calibrated:
3 Point Traceable
3-point UKAS
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Calibrated unitSize
17873288 3-point UKAS Each
17813058 3 Point Traceable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17873288 Supplier Brannan™ Supplier No. 17/102/2 + 49/503/0
Add to basket
Add to basket
This item is not returnable. View return policy

Precision test oven thermometers sealed into a 30 mL polypropylene bottle containing a heat transfer medium, ideally suited to monitor the temperature of samples in a controlled storage. Complete with traceable calibration.

  • Non-hazardous LO-tox™ filling
  • Temperature measurement unaffected by short term fluctuations
  • Temperature range: +20°C to +130°C
  • Supplied with calibration and statement of accuracy
  • Made in UK

Specifications

Bottle Fill Aluminium oxide crystals
Bottle Size 30 mL
Calibration Points 30°C, 80°C, 120°C
Certifications/Compliance ISO 17025 UKAS
For Use With (Application) Oven Batch Sampling
Height (English) 5.71 in.
Height (Metric) 14.5 cm
Includes Polypropylene bottle and glass thermometer
Length (Metric) 14.5 cm
Material Glass
Thermometer Fill Aluminium oxide crystals
Weight (English) 3.2099 oz.
Weight (Metric) 91 g
Width (English) 0.14 in.
Max. Temperature (English) 266°F
Max. Temperature (Metric) +130°C
Min. Temperature (English) 68°F
Min. Temperature (Metric) +20°C
Temperature Range (English) 68°F to 266°F
Temperature Range (Metric) +20°C to +130°C
Calibrated 3-point UKAS
Battery Type None
Accuracy ± 2°C
Subdivision 1.0°C
Display None
Width (Metric) 3.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.