missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brannan™ Batch Sampling Oven Thermometer with Calibration Certificate
Click to view available options
Calibrated:
3 Point Traceable
3-point UKAS
Unit Size:
Each
Description
- Non-hazardous LO-tox™ filling
- Temperature measurement unaffected by short term fluctuations
- Temperature range: +20°C to +130°C
- Supplied with calibration and statement of accuracy
- Made in UK
Specifications
Specifications
| Bottle Fill | Aluminium oxide crystals |
| Bottle Size | 30 mL |
| Calibration Points | 35°C, 50°C, 90°C |
| Certifications/Compliance | UK National Standards ISO 9001 |
| For Use With (Application) | Oven Batch Sampling |
| Height (English) | 5.71 in. |
| Height (Metric) | 14.5 cm |
| Includes | Polypropylene bottle and glass thermometer |
| Length (Metric) | 14.5 cm |
| Material | Glass |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction