missing translation for 'onlineSavingsMsg'
Learn More

Brannan™ Batch Sampling Incubator Thermometer with Calibration Certificate

Product Code. 17863118
Click to view available options
Calibrated:
3 Point Traceable
3-point UKAS
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17863118

Brand: Brannan™ 22/105/2 + 49/101/0

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Precision test incubator thermometers sealed into a 30 mL polypropylene bottle containing a heat transfer medium, ideally suited to monitor the temperature of samples in a controlled storage. Complete with calibration.

  • Non hazardous LO-tox™ filling
  • Bottle should be filled with monpropylene glycol or other heat transfer medium before use
  • Temperature measurement unaffected by short term fluctuations
  • Temperature range: 18°C to 50°C
  • Supplied with calibration and statement of accuracy
  • Made in UK

Specifications

Bottle Fill Customer to fill with monopropylene glycol or other heat transfer medium before use
Calibration Points 20°C, 35°C, 50°C
Certifications/Compliance UK National Standards ISO 9001
For Use With (Application) Monitor temperature of samples in a controlled storage
Height (English) 5.71 in.
Height (Metric) 14.5 cm
Includes Polypropylene bottle and glass thermometer
Length (Metric) 14.5 cm
Material Glass
Thermometer Fill No
Weight (English) 1.9577 oz.
Weight (Metric) 55.5 g
Width (English) 0.14 in.
Max. Temperature (English) 122°F
Max. Temperature (Metric) +50°C
Min. Temperature (English) 64°F
Min. Temperature (Metric) +18°C
Temperature Range (English) 64°F to 122°F
Temperature Range (Metric) +18°C to +50°C
Calibrated 3 Point Traceable
For Use With (Equipment) Incubator
Battery Type None
Accuracy ± 0.5°C
Subdivision 1.0°C
Display None
Width (Metric) 3.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.