missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bat Immunoglobulin Antibody (BT1-4F10), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23483H
This item is not returnable.
View return policy
Description
Bat Immunoglobulin Monoclonal specifically detects Bat Immunoglobulin in Bat samples. It is validated for Western Blot, ELISA.
Specifications
| Bat Immunoglobulin | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| Microchiropteran bat (Eptesicus fuscus) spleen cells. | |
| 0.1 mL | |
| Primary | |
| Bat | |
| Purified |
| Western Blot, ELISA | |
| BT1-4F10 | |
| Western Blot, ELISA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Immunoglobulin L (Light) chain. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction