missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bassoon Antibody (SAP7F407), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB120-13249-0.2MG
This item is not returnable.
View return policy
Description
Bassoon Monoclonal specifically detects Bassoon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Bassoon | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:400, Immunoprecipitation 2.5 ug, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen | |
| O88778 | |
| BSN | |
| Recombinant rat Bassoon. | |
| Protein G purified | |
| RUO | |
| 8927 | |
| Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| SAP7F407 | |
| Unconjugated | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| bassoon (presynaptic cytomatrix protein) | |
| Mouse | |
| 400 kDa | |
| 0.2 mg | |
| Primary | |
| This affinity purified detects an ∼400 kDa protein, corresponding to the apparent molecular mass of Bassoon on SDSPAGE immunoblots, in samples from mouse and rat origins. Additional bands between 97 and 400 kDa may also be detected. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction