missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bassoon Antibody (SAP7F407), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-13249H
This item is not returnable.
View return policy
Description
Bassoon Monoclonal antibody specifically detects Bassoon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| Bassoon | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| SAP7F407 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| bassoon (presynaptic cytomatrix protein) | |
| Recombinant rat Bassoon. | |
| 0.2 mL | |
| Neuronal Cell Markers, Neuroscience | |
| 8927 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction