missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BASP1 Polyclonal Antibody
BASP1 Polyclonal Antibody
Brand: Invitrogen PA545048
This item is not returnable.
View return policy
Description
BASP1 Polyclonal Antibody for Western Blot
Specifications
| BASP1 | |
| Polyclonal | |
| Unconjugated | |
| BASP1 | |
| 22 kDa neuronal tissue-enriched acidic protein; 2610024P12Rik; BASP1; brain abundant membrane attached signal protein 1; brain abundant, membrane attached signal protein 1; brain acid soluble protein 1; brain acidic membrane protein; brain acid-soluble protein 1; CAP23; CAP-23; Ckap3; cytoskeleton associated protein 3; MGC8555; NAP22; NAP-22; neuronal axonal membrane protein NAP-22; neuronal tissue-enriched acidic protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10409 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P80723 | |
| BASP1 | |
| Synthetic peptide directed towards the C-terminal of human BASP1 (aa 134-183). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction