missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BASP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £255.00
Specifications
| Antigen | BASP1 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18669615
|
Novus Biologicals
NBP2-99048-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673536
|
Novus Biologicals
NBP2-99048-50ul |
50 μg |
£178.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
BASP1 Polyclonal antibody specifically detects BASP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Especificaciones
| BASP1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| PBS (pH 7.0) | |
| 10409 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| brain abundant, membrane attached signal protein 1,22 kDa neuronal tissue-enriched acidic protein, brain acid-soluble protein 1, CAP-23, CAP23brain acid soluble protein 1, NAP-22, NAP22MGC8555, Neuronal axonal membrane protein NAP-22, neuronal tissue-enriched acidic protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human BASP1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto