missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BASP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | BASP1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
BASP1 Polyclonal specifically detects BASP1 in Human, Rat samples. It is validated for Western Blot.Specifications
| BASP1 | |
| Polyclonal | |
| Rabbit | |
| P80723 | |
| 10409 | |
| Synthetic peptides corresponding to BASP1 (brain abundant, membrane attached signal protein 1) The peptide sequence was selected from the C terminal of BASP1. Peptide sequence APAAGEEPSKEEGEPKKTEAPAAPAAQETKSDGAPASDSKPGSSEAAPSS. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| brain abundant, membrane attached signal protein 1,22 kDa neuronal tissue-enriched acidic protein, brain acid-soluble protein 1, CAP-23, CAP23brain acid soluble protein 1, NAP-22, NAP22MGC8555, Neuronal axonal membrane protein NAP-22, neuronal tissue-enriched acidic protein | |
| BASP1 | |
| IgG | |
| 23 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title