Learn More
Buerkle™ Basket for Sample Bottles Vampire
Product Code. 16190891 Shop All Bürkle Products
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. Material unitSize
16190891 Stainless Steel Each
1 options
Product Code. 16190891 Supplier Buerkle™ Supplier No. 53022101
Basket for use with Buerkle™ Vampire Sampler. Suitable for 1000 ml bottles.
Diameter 100 mm.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Bottle Holder
Material Stainless Steel
Autoclavable Autoclavable
Disposable Reusable
Sterility Non-sterile
For Use With (Application) For use with Buerkle Vampire Sampler
For Use With (Equipment) Bottle holder