missing translation for 'onlineSavingsMsg'
Learn More

Basic Red 46, Technical Grade, TRC

Product Code. 17984005
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17984005 25 mg 25mg
17941325 50 mg 50mg
17912195 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17984005 Supplier Toronto Research Chemicals Supplier No. TRCB11873325MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12221-69-1
Molecular Formula C18 H21 N6 . Br
Molecular Weight (g/mol) 401.3
Synonym 1,4-Dimethyl-5-[[4-[methyl(phenylmethyl)amino]phenyl]azo]-1H-1,2,4-triazolium Bromide, Anilan Red GRL, Astrazon Red FBL, Astrazon Red FBLN, Asucryl Red GRL, Basic Red 113, Basic Red 46, C.I. 110825, C.I. Basic Red 113, C.I. Basic Red 46, Cationic Red GRL, Cationic Red SD-GRL, Cationic Red X-GRL, Diacryl Red RL-N, Disperse Cationic Red SD-GRL, Dystar Red FBL, Estrol Red N-GSL, Kayacryl Red GRL, Kayacryl Red GRL-ED, Kayacryl Red GRL-N, Maxilon Red GRL, Maxilon Red GRL Pearls, R X-GRL, Red X-GRL, Sandocryl Red B-RLE, Sevron Fast Red GRL 200%, Synacril Red G
IUPAC Name N-benzyl-4-[(E)-(2,4-dimethyl-1,2,4-triazol-4-ium-3-yl)azo]-N-methyl-aniline;bromide
SMILES [Br-].CN(Cc1ccccc1)c2ccc(cc2)N=Nc3[n+](C)cnn3C

Specifications

Analyte or Component Names Basic Red 46
Shipping Condition Room Temperature
InChI Formula InChI=1S/C18H21N6.BrH/c1-22(13-15-7-5-4-6-8-15)17-11-9-16(10-12-17)20-21-18-23(2)14-19-24(18)3;/h4-12,14H,13H2,1-3H3;1H/q+1;/p-1
Solution Type Neat
Industry Type Forensics
Quantity 25 mg
Formula Weight 400.1011
Chemical Name or Material Basic Red 46
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.