Learn More
Tork Basic Paper W6, Wiping Paper Towel Roll
Product Code. 30243703 Shop All Tork Products
Change view
Click to view available options
Color:
Blue
White
Unit Size:
6 rolls
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30243703 Blue 6 rolls
30243782 White 6 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30243703 Supplier Tork Supplier No. 130008
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Tork Paper Wiper is designed for optimal dispensing from the Tork Washstation Dispenser System. HACCP International certification ensures that this product is safe for use in food processing and food preparation areas.
  • Efficiently clean, even in high humidity and wet environments, with this specially designed wiping paper.
  • Ensure you have high-quality wiping paper, thanks to these food contact certified rolls.
  • Making your environment HACCP compliant is made easier with these HACCP International certified rolls.
  • Improve food production safety, thanks to the high visibility and traceability of this paper roll.
  • 6 x 250 m roll
TRUSTED_SUSTAINABILITY
Specifications
Product Type Wiping Paper
Length (English) 273.4 yd.
Length (Metric) 250 m
Width (English) 7.68 in.
Width (Metric) 19.5 cm
Autoclavable Not Autoclavable
Color Blue
Packaging Quantity 1 Roll
Ply 1-ply