missing translation for 'onlineSavingsMsg'
Learn More

Basic Green 4 (Technical Grade), TRC

Product Code. 30012805
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012805 5 g 5g
30003786 10 g 10g
30004235 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012805 Supplier Toronto Research Chemicals Supplier No. TRCB1276335G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 569-64-2
Molecular Formula C23 H25 N2 . Cl
Molecular Weight (g/mol) 364.91
Synonym Methanaminium, N-[4-[[4-(dimethylamino)phenyl]phenylmethylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-, chloride (1:1), C.I. Basic Green 4 (8CI), Methanaminium, N-[4-[[4-(dimethylamino)phenyl]phenylmethylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-, chloride (9CI), Victoria Green WB (6CI), 65: PN: WO2014160752 SEQID: 215 claimed sequence, ADC Malachite Green Crystals, Acryl Brilliant Green B, Aizen Malachite Green, Aizen Malachite Green Crystals, Aizen malachite green liquid, Aniline green, Astra Malachite Green, Astra Malachite Green B, Astra Malachite Green BXX, Astrazon Green M, Atlantic Malachite Green, Basacryl Green X-BF, Basic Green 4, Basic Light Green G 4, Basonyl Green 830, Basonyl Green NB 832, Benzal Green, Benzaldehyde green, Bronze Green Toner A 8002, Burma Green B, C.I. 42000, Calcozine Green V, China Green, Diabasic Malachite Green, Diamond Green B, Diamond Green B extra, Diamond Green BX, Diamond Green P Extra, Green MX, Green Malaquite, Grenoble Green, Hidaco Malachite Green Base, Hidaco Malachite Green LC, Hidaco Malachite Green SC, LC 6220, Light Green N, Lincoln Green Toner B 15-2900, MKG, Malachite Green 4, Malachite Green A, Malachite Green AN, Malachite Green B, Malachite Green CP, Malachite Green Crystals, Malachite Green Crystals BPC, Malachite Green J 3E, Malachite Green Powder, Malachite Green WS, Malachite Green XLS, Malachite Lake Green A, Malachite green, Malachite green chloride, Mitsui Malachite Green, New Victoria Green Extra I, New Victoria Green Extra II, New Victoria Green Extra O, Oji Malachite Green, Solid Green Crystals O, Solid Green O, Super Ick Cure, Tertrophene Green M, Tokyo Aniline Malachite Green, Verona Basic Green M, Victoria Green, Victoria Green (basic dye), Victoria Green B, Victoria Green S, Victoria Green WPB
IUPAC Name [4-[[4-(dimethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES [Cl-].CN(C)c1ccc(cc1)C(=C2C=CC(=[N+](C)C)C=C2)c3ccccc3

Specifications

Quantity 5 g
InChI Formula InChI=1S/C23H25N2.ClH/c1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4;/h5-17H,1-4H3;1H/q+1;/p-1
Formula Weight 364.1706
Chemical Name or Material Malachite Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.